Learn More
His-tag Rabbit, Polyclonal, GenScript™
Rabbit Polyclonal Antibody
Supplier: Genscript Corporation A0017440

Description
Reconstitute the lyophilized powder with deionized water (or equivalent) to an antibody concentration of 0.5 mg/mL. His-tag Antibody, pAb, Rabbit recognizes His tagged proteins in transfected or transformed cells. Short-sequence epitope tags are common in the study of protein expression in several different expression systems. In particular, the His tag on the expressed recombinant proteins allows for efficient coupling to Ni2+ affinity resins and purification by single step chromatography.
His-tag Polyclonal antibody specifically detects His-tag in Tag samples. It is validated for ELISA, Western BlotSpecifications
His-tag | |
Polyclonal | |
PBS with 0.02% sodium azide | |
Rabbit | |
Affinity chromatography | |
RUO | |
-20°C or -80°C if preferred | |
Lyophilized | |
Tag |
ELISA, Western Blot | |
Unconjugated | |
6His tag; 6X His; His Tag; Histidine Tag | |
His epitope tag peptide HHHHHH conjugated to KLH | |
40 μg | |
Primary | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.