Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD148 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578139
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578139 Supplier Invitrogen™ Supplier No. PA578139
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: K562, K562 membrane extract Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CD148 is a highly glycosylated transmembrane protein of up to 250 kDa, containing eight fibronectin type III domains in the extracellular region and one tyrosine phosphatase domain in the cytoplasmic region. Expression is found broadly on fibroblasts, epithelial cells, lymphocytes (B, T and NK cells), and myeloid cells (monocytes and granulocytes). CD148 can dephosphorylate proteins such as LAT and PLC-gamma, and is involved in the downregulation of T cell activation. Conversely, CD148 expression can be upregulated following T cell activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD148
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene PTPRJ
Gene Accession No. Q12913
Gene Alias AI450271; BET; Byp; CD148; CD148 antigen; colon tumor susceptibility 1; density enhanced phosphatase-1; density-enhanced phosphatase 1; DEP1; DEP-1; HPTP beta-like tyrosine phosphatase; HPTP eta; HPTPeta; human density enhanced phosphatase-1; protein tyrosine phosphatase, receptor type J; protein tyrosine phosphatase, receptor type, J; protein tyrosine phosphatase, receptor type, J polypeptide; protein-tyrosine phosphatase eta; Protein-tyrosine phosphatase receptor type J; Ptpb2; PTPbeta2; Ptprj; Receptor-type tyrosine-protein phosphatase eta; R-PTP-ETA; RPTPJ; R-PTP-J; Scc1; Scc-1; Susceptibility to colon cancer 1; susceptibility to colon cancer 1, mouse, homolog of
Gene Symbols PTPRJ
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human PTPRJ. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5795
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.