Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiScreen™ Capto™ S, Capto™ Q and Capto™ DEAE Prepacked IEX Columns

Catalog No. 45002530 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
HiScreen Capto DEAE IEX Column
HiScreen Capto Q IEX column
HiScreen Capto S IEX column
Ligand Type:
Diethylaminoethyl
Quaternary Amine
Sulphonate group
pH Range:
2 to 12
2 to 9
4 to 12
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Ligand Type pH Range
45-002-530 HiScreen Capto S IEX column Sulphonate group 4 to 12
45-002-529 HiScreen Capto Q IEX column Quaternary Amine 2 to 12
45-002-533 HiScreen Capto DEAE IEX Column Diethylaminoethyl 2 to 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-002-530 Supplier Cytiva Supplier No. 28926979
Add to Cart
Add to Cart

Designed for method optimization and parameter screening

  • Fast, efficient, and cost-effective capture and intermediate protein purification
  • Ready-to-use columns for convenient process development optimization
  • Highly cross-linked agarose with dextran surface extender matrix
  • Bed height of 10cm is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
  • Combines high capacity with high flow velocity and low backpressure to reduce process cycle times and increase productivity
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity

Capto S: Strong cation (sulfonate group) exchange BioProcess™ medium
Capto Q: Strong anion (quartenary amine group) exchange BioProcess medium
Capto DEAE: Weak anion (diethylaminoethyl group) exchange BioProcess medium

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M Acetic Acid, 1M NaOH, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
Product Type HiScreen Capto S IEX column
Bed Volume 4.7mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.7mm
Quantity 1 x 4.7mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate <700 cm/hr.
Material Polypropylene
Ligand Type Sulphonate group
Max. Pressure 3 bar
pH Range 4 to 12
Particle Size 45 to 165μm
Support Type Highly cross-linked Agarose with Dextran surface extender
Content And Storage 4° to 30°C
Column Type Ion Exchange Column
For Use With (Application) Modern large-scale manufacturers for fast, efficient and cost-effective capture and intermediate Protein purification, optimization and parameter screening
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.