Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HisG Epitope Tag Monoclonal Antibody, HRP
HisG Epitope Tag Monoclonal Antibody, HRP
Supplier: LIFE TECHNOLOGIES R94125

Description

Specifications
HisG Epitope Tag | |
Monoclonal | |
PBS with no preservative | |
Mouse | |
purified | |
RUO | |
Tag | |
Antibody | |
IgG2a |
ELISA, Western Blot | |
HRP | |
6xHis-Gly; 6xHis-Glycine; His G; His Tag; Histidine Tag | |
6x His synthetic peptide: -His-His-His-His-His-His-Gly. | |
50 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction