Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HISPPD1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB266203 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB266203 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB266203 Supplier Novus Biologicals Supplier No. NBP232155AF532
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HISPPD1 Polyclonal antibody specifically detects HISPPD1 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen HISPPD1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias diphosphoinositol pentakisphosphate kinase 2FLJ21506, EC 2.7.4.21, EC 2.7.4.24, HISPPD1histidine acid phosphatase domain containing 1, Histidine acid phosphatase domain-containing protein 1, hsVIP2, inositol heptaphosphate kinase 2, inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2, InsP6 and PP-IP5 kinase 2, KIAA0433FLJ23463, VIP1 homolog 2, VIP2IP7K2
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 925 to 975 of human Diphosphoinositol Pentakisphosphate Kinase 2 using the numbering given in entry BAA24863.2 (GeneID 23262).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 23262
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.