Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HISPPD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216836
Description
HISPPD1 Polyclonal specifically detects HISPPD1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HISPPD1 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
diphosphoinositol pentakisphosphate kinase 2FLJ21506, EC 2.7.4.21, EC 2.7.4.24, HISPPD1histidine acid phosphatase domain containing 1, Histidine acid phosphatase domain-containing protein 1, hsVIP2, inositol heptaphosphate kinase 2, inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2, InsP6 and PP-IP5 kinase 2, KIAA0433FLJ23463, VIP1 homolog 2, VIP2IP7K2 | |
Rabbit | |
140 kDa | |
0.1 mL | |
Protein Phosphatase | |
23262 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
O43314 | |
PPIP5K2 | |
Recombinant protein encompassing a sequence within the N-terminus region of human HISPPD1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (99%), Xenopus laevis (94%), Bovine (99%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction