Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HISPPD1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | HISPPD1 |
---|---|
Concentration | 0.24 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5168
![]() ![]() |
Proteintech Group Inc
260181AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Inositol phosphates.Specifications
HISPPD1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O43314 | |
23262 | |
HISPPD1 Fusion Protein Ag23374 | |
Primary | |
-20°C | |
PPIP5K2 |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HISPPD1, hsVIP2, InsP6 and PP IP5 kinase 2, IP7K2, KIAA0433, PPIP5K2, VIP1 homolog 2, VIP2 | |
PPIP5K2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title