Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HISPPD2A Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP244293AF700
Description
HISPPD2A Polyclonal antibody specifically detects HISPPD2A in Human samples. It is validated for Western BlotSpecifications
HISPPD2A | |
Polyclonal | |
Western Blot | |
diphosphoinositol pentakisphosphate kinase 1, histidine acid phosphatase domain-containing protein 2A, inositol pyrophosphate synthase 1, VIP1 | |
The epitope recognized by this antibody maps to a region between residue 1383 to 1433 of human Inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 1 using the numbering given in entry Q6PFW1.1 (GeneID 9677). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
9677 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction