Learn More
Thermo Scientific™ HisPur™ Ni-NTA Magnetic Beads, 2 mL PROMO
Description
The blocked magnetic bead surface is coated with anti–HA antibody, a highly specific mouse IgG1 monoclonal antibody that recognizes the HA-epitope tag (YPYDVPDYA) derived from the human influenza hemagglutinin (HA) protein. The Pierce Anti-HA Magnetic Beads can be used manually with a magnetic stand, as well as with automated platforms, such as the Thermo Scientific™ KingFisher Instruments, for high-throughput workflows.
Highlights:
Specific – highly specific anti–HA monoclonal antibody enables high–yield and high-purity immunoprecipitation (IP)
Fast – optimized protocol is completed in approximately 1 hour
Low nonspecific binding – stable, pre–blocked beads provide highly purified HA-tagged proteins
Versatile – beads are compatible with manual and automated workflows (e.g., Thermo Scientific™ KingFisher Instruments)
Specifications
Specifications
| Content And Storage | Store at 4°C |
| Form | Liquid |
| Product Line | HisPur™ |
| Quantity | 2 mL |
| Protein Form | Recombinant |
| Target | His |
| Type | Magnetic Affinity Bead |
| Ligand Type | Nickel-NTA |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.