Learn More
Thermo Scientific™ HisPur™ Ni-NTA Resin, 100 mL PROMO
Description
The specially prepared nickel-NTA support consists of beaded agarose derivatized with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni2+). The immobilized metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.
HisPur Ni-NTA Resin is available in several package sizes of resin slurry, three sizes of centrifuge-ready columns, complete purification kits, two sizes of FPLC-ready chromatography cartridges, and 96-well filter plates for high through-put needs. With these options, high-yield His-tag protein purification is possible at nearly any scale.
Highlights:
- High capacity – bind up to 60mg of 6xHis-tagged protein per milliliter of resin
- Versatile – purify proteins using native or denaturing conditions
- Compatible – use with Thermo Scientific Cell Lysis Reagents and a variety of buffer additives
- Flexible – available in multiple formats including bulk resin, spin columns and chromatography cartridges
- Cost-effective – reuse the same batch of resin at least five times
- Easy to use – pre-formulated buffers available for kit formats
Available Formats:
- Resin Slurries – crosslinked 6% beaded agarose; 10 mL, 100 mL, 500 mL bottles
- Spin Columns – 0.2 mL (microcentrifuge), 1 mL, 3 mL columns
- Purification Kits – complete kits in all three column sizes
- Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
- Bulk and Custom – let us know your needs for bulk HisPur Ni-NTA Resin
Includes:
Kits contain HisPur Ni-NTA Columns (store at 4°C)
Specifications
Specifications
Capacity (Metric) | Up to 60 mg of 6xHis-Tagged Protein Binding per mL Resin |
Content And Storage | Store at 4°C |
Form | Liquid Suspension |
Product Line | HisPur™ |
Quantity | 100 mL |
Target | His-Tagged Protein |
Type | Resin |
Stationary Phase | Divalent Nickel, Metal Chelate |
Format | Bottle |
Column Type | Affinity, Agarose Resin |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.