Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HisPur™ Ni-NTA Spin Purification Kit, 3 mL PROMO

Catalog No. PI88229PM
Change view
Click to view available options
:
Thermo Scientific™ HisPur™ Ni-NTA Spin Purification Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PI88229PM Thermo Scientific™ HisPur™ Ni-NTA Spin Purification Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88229PM Supplier Thermo Scientific™ Supplier No. 88229

Purify His-tagged recombinant fusion proteins using high-capacity nickel-NTA (nitrilotriacetic acid) metal-chelate beads, spin columns and plates.

The specially prepared nickel-NTA support consists of beaded agarose derivatized with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni2+). The immobilized metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.

HisPur Ni-NTA Resin is available in several package sizes of resin slurry, three sizes of centrifuge-ready columns, complete purification kits, two sizes of FPLC-ready chromatography cartridges, and 96-well filter plates for high through-put needs. With these options, high-yield His-tag protein purification is possible at nearly any scale.

Highlights:

  • High capacity – bind up to 60mg of 6xHis-tagged protein per milliliter of resin

  • Versatile – purify proteins using native or denaturing conditions

  • Compatible – use with Thermo Scientific Cell Lysis Reagents and a variety of buffer additives

  • Flexible – available in multiple formats including bulk resin, spin columns and chromatography cartridges

  • Cost-effective – reuse the same batch of resin at least five times

  • Easy to use – pre-formulated buffers available for kit formats

Available Formats:

  • Resin Slurries – crosslinked 6% beaded agarose; 10 mL, 100 mL, 500 mL bottles
  • Spin Columns – 0.2 mL (microcentrifuge), 1 mL, 3 mL columns
  • Purification Kits – complete kits in all three column sizes
  • Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
  • Bulk and Custom – let us know your needs for bulk HisPur Ni-NTA Resin

Includes:

Kits contain HisPur Ni-NTA Columns

Specifications

Capacity (Metric) 1 mL/Column
Content And Storage Sufficient For: Purifying up to 180 mg of His-tagged protein per column, HisPur Ni-NTA Spin Columns, 3 mL, 5 Columns, Phosphate-buffered Saline (10X), 30 mL, Imidazole (2M), 25 mL, Column Plugs, 5 Plugs, Store at 4°C.
Product Line HisPur
Quantity 5 Column Kit
Target His-Tagged Protein
Type Spin Column Kit
Stationary Phase Divalent Nickel, Metal Chelate
Sufficient For 180 mg of His-Tagged Protein per Column
Format Spin Column
Column Type Affinity, Agarose Resin

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.