Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIST1H2AB Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305881APC
Description
HIST1H2AB Polyclonal antibody specifically detects HIST1H2AB in Human samples. It is validated for Western BlotSpecifications
HIST1H2AB | |
Polyclonal | |
Western Blot | |
histone H2A type 1-B/E, H2A/m, H2AFM, histone cluster 1, H2ab | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human HIST1H2AB. | |
0.1 mL | |
Epigenetics | |
8335 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction