Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIST1H2AG Antibody (CL5041), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP26115225UL
Description
HIST1H2AG Monoclonal specifically detects HIST1H2AG in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HIST1H2AG | |
CL5041 | |
Immunohistochemistry-Paraffin 1:200 - 1:500 | |
H2A histone family, member P, H2A.1, H2A.1b, H2A/p, H2AFC, H2AFD, H2AFI, H2AFN, H2AFP, HIST1H2AI, HIST1H2AK, HIST1H2AL, HIST1H2AM, histone 1, H2ag, histone cluster 1, H2ag, histone H2A type 1, Histone H2A/p, pH2A/f | |
Mouse | |
Protein A purified | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Purified |
Monoclonal | |
Unconjugated | |
PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide | |
HIST1H2AG | |
This antibody was developed against a synthetic peptide corresponding to the following amino acid sequence:RSSRAGLQFPVGRVHR | |
25 μL | |
8969 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction