Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIST1H2AG Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA524822

Catalog No. PIPA524822


Only null left
Add to Cart

Description

Description

This antibody is predicted to react with bovine, mouse and rat based on sequence homology.

Histones are nuclear proteins that play an important role in maintaining the structure of the chromosome in eukaryotes. The nucleosome is made up of around 146bp of DNA, which enfolds around a histone octomer unit comprising of four major histones, H2A, H2B, H3 and H4. An important role of H2A is to ensure proper assembly or functioning of the centrometre. They also play an important role in gene regulation. The protein can be modified by acetylation and phosphorylation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HIST1H2AG
Polyclonal
Unconjugated
H2AC11
H2A histone family, member P; H2A.1; H2A.1b; H2A/d; H2A/p; H2AC11; H2AC13; H2AC15; H2AC16; H2AC17; H2AFC; H2AFD; H2AFI; H2AFN; H2AFP; H2AG; H2ak; HIST1H2AG; HIST1H2AI; HIST1H2AJ; HIST1H2AK; HIST1H2AL; HIST1H2AM; histone 1, H2ag; histone cluster 1, H2ag; histone H2A type 1; histone H2A/e; histone H2A/p; Histone H2A/ptl; pH2A/f
Rabbit
Protein A, Antigen affinity chromatography
RUO
8329, 8969
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ChIP Assay, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P0C0S8
H2AC11, HIST1H2AG
KLH conjugated synthetic peptide between 103-130 amino acids from the C-terminal region of human HIST1H2AL.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.