Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIST1H2AH Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | HIST1H2AH |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
HIST1H2AH Polyclonal specifically detects HIST1H2AH in Human samples. It is validated for Western Blot.Specifications
HIST1H2AH | |
Polyclonal | |
Rabbit | |
NP_542163 | |
85235 | |
The immunogen for this antibody is HIST1H2AH. Peptide sequence: IIPRHLQLAIRNDEELNKLLGKVTIAQGGVLPNIQAVLLPKKTESHHKAK | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
histone cluster 1, H2ah, histone H2A type 1-H | |
HIST1H2AH | |
IgG | |
14 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title