Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIST1H2BA Antibody (OTI1C9), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272479C
Description
HIST1H2BA Monoclonal specifically detects HIST1H2BA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
HIST1H2BA | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HIST1H2BA | |
Recombinant protein fragment corresponding to amino acids 2-127 of human HIST1H2BA (NP_733759) produced in E.coli. | |
0.1 mL | |
255626.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1C9 | |
Western Blot, Immunohistochemistry | |
histone H2B type 1-A, bA317E16.3, H2BFU, histone cluster 1, H2ba, STBP, TSH2B | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction