Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIST1H2BA Monoclonal Antibody (OTI1C9)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526966
Description
HIST1H2BA Monoclonal Antibody for Western Blot, IHC (P)
Variant histone specifically required to direct the transformation of dissociating nucleosomes to protamine in male germ cells. Entirely replaces classical histone H2B prior nucleosome to protamine transition and probably acts as a nucleosome dissociating factor that creates a more dynamic chromatin, facilitating the large-scale exchange of histones. Also expressed maternally and is present in the female pronucleus, suggesting a similar role in protamine replacement by nucleosomes at fertilization. Also found in fat cells, its function and the presence of post-translational modifications specific to such cells are still unclear. Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Specifications
| HIST1H2BA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96A08 | |
| H2bc1 | |
| Human recombinant protein fragment corresponding to amino acids 2-127 of HIST1H2BA produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C9 | |
| Unconjugated | |
| H2bc1 | |
| bA317E16.3; H2B histone family, member U, (testis-specific); H2BFU; HIST1H2BA; histone 1, H2ba; histone cluster 1, H2ba; histone H2B type 1-A; Histone H2B, testis; STBP; testis-specific histone H2B; TH2B; TSH2B; TSH2B.1; TSH2B1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 255626 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction