Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIST1H2BB Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117474
Description
This antibody has been tested in direct-ELISA.
Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Specifications
| HIST1H2BB | |
| Polyclonal | |
| Unconjugated | |
| H2BC3 | |
| H2B histone family, member F; H2B.1; H2B/f; h2B-143; H2B-clustered histone 3; H2BFF; HIST1H2BB; histone 1, H2bb; histone cluster 1, H2bb; histone H2B type 1-B; Histone H2B.1; histone H2B.f | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3018, 319178 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P33778, Q64475 | |
| H2BC3, HIST1H2BB | |
| Peptide sequence around site of Lys (16) derived from Human Histone H2B type 1-B. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction