Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIST1H2BC Antibody (CL5048), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | HIST1H2BC |
---|---|
Clone | CL5048 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
HIST1H2BC Monoclonal specifically detects HIST1H2BC in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HIST1H2BC | |
Monoclonal | |
Purified | |
Human, Mouse, Rat | |
8347 | |
This antibody was developed against a synthetic peptide corresponding to the following amino acid sequence:VSEGTKAVTKYTSSK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL5048 | |
Unconjugated | |
Mouse | |
H2B histone family, member K, H2B/a, H2B/g, H2B/kH2B/h, H2BFA, H2BFG, H2BFH, H2BFKH2B/l, H2BFL, HIST1H2BC, HIST1H2BE, HIST1H2BF, HIST1H2BG, histone 1, H2bi, histone cluster 1, H2bi, histone H2B type 1-C/E/F/G/I, Histone H2B.1 A, Histone H2B.a, Histone H2B.g, Histone H2B.h, Histone H2B.k, Histone H2B.l | |
HIST1H2BC | |
IgG2b | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title