Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIST1H2BK Recombinant Rabbit Monoclonal Antibody (4G10)
GREENER_CHOICE

Catalog No. PIMA533155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533155 Supplier Invitrogen™ Supplier No. MA533155
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

HIST1H2BK Recombinant Monoclonal Antibody for Western Blot, IHC (P), ELISA

Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIST1H2BK
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 4G10
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene H2BC12
Gene Accession No. O60814
Gene Alias H2B histone family, member T; H2B K; H2B/S; H2BFAiii; H2BFT; H2BK; HIRA-interacting protein 1; HIRIP1; HIST1H2BK; histone 1, H2bk; histone cluster 1, H2bk; histone family member; histone H2B type 1-K
Gene Symbols H2BC12
Host Species Rabbit
Immunogen Synthesized HIST1H2BK (100-126aa).
Purification Method Protein G, Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85236
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.