Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIST1H2BM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549358
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549358 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549358 Supplier Invitrogen™ Supplier No. PA549358
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HIST1H2BM Polyclonal Antibody for Western Blot

Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIST1H2BM
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene H2BC14
Gene Accession No. Q99879
Gene Alias dJ160A22.3; H2B histone family, member E; H2B/e; H2BFE; HIST1H2BM; histone 1, H2bm; histone cluster 1, H2bm; histone H2B type 1-M; Histone H2B.e; Histone H2Be
Gene Symbols H2BC14
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 10-44 amino acids from the N-terminal region of human HIST1H2BM.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8342
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.