Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST1H3A, anti-Human, Biotin, Clone:, Abnova™

Catalog No. 89998428 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-428 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-428 Supplier Abnova Corporation Supplier No. MAB15852
Add to Cart
Add to Cart

Monoclonal Antibody

Specifications

Antigen HIST1H3A
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Immunofluorescence
Classification Monoclonal
Conjugate Biotin
Dilution ChIP (0.2-1.8 ug/ChIP) Immunofluorescence (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBSE (0.2% sucrose).
Gene histone cluster 1, H3a
Gene Alias H3/A/H3FA
Gene Symbols HIST1H3A
Immunogen Recombinant antibody corresponding to Histone H3, trimethylated at lysine 9.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8350
Includes negative control
Target Species Fruitfly, Human, Mouse, Yeast
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.