Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST1H3A, Formyl Lys122 Antibody (32C1), Novus Biologicals™
SDP

Catalog No. NB297829 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297829 100 μL
NB297828 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297829 Supplier Novus Biologicals Supplier No. NBP326574100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HIST1H3A Monoclonal antibody specifically detects HIST1H3A in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen HIST1H3A
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 32C1
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias H3 histone family, member A, H3/A, H3FAHIST1H3E, H3FB, H3FC, H3FD, H3FF, H3FH, H3FI, H3FJ, H3FK, H3FL, HIST1H3B, HIST1H3C, HIST1H3D, HIST1H3F, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J, histone 1, H3a, histone cluster 1, H3a, histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l
Host Species Rabbit
Immunogen A synthesized peptide derived from Human HIST1H3A. The HIST1H3A [Formyl Lys122] Recombinant Monoclonal Antibody was raised against the peptide IMP-(Fo)K-DI formylated at K122. [UniProt P68431]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cellular Markers, Core ESC Like Genes, Oncogenes, Protein Kinase, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8350
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.