Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST1H3A, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89998410 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-410 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-410 Supplier Abnova Corporation Supplier No. PAB31327
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen HIST1H3A
Applications Dot Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA (1:5000-10000) Dot Blot (1:20000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide, 0.05% proclin 300).
Gene histone cluster 1, H3a
Gene Alias H3/A/H3FA
Gene Symbols HIST1H3A
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to Histone H3, phosphorylated at threonine 45.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8350
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.