Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST1H3A Rabbit anti-Human, Unconjugated, Clone: RM339, Abnova™

Catalog No. 89339182 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-182 Supplier Abnova Corporation Supplier No. MAB21965
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

HIST1H3A Monoclonal antibody specifically detects HIST1H3A in Human samples. It is validated for WB, Dot-Pep, M-PCR.

Specifications

Antigen HIST1H3A
Applications Western Blot, Dot Blot, Multiplex
Classification Monoclonal
Clone RM339
Conjugate Unconjugated
Description histone cluster 1, H3a
Gene HIST1H3A
Gene Alias H3/A, H3FA
Host Species Rabbit
Immunogen An crotonyl-peptide corresponding to HIST1H3A (crotonyl K9)
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 8350
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.