Learn More
HIST1H3A Rabbit anti-Human, Unconjugated, Clone: RM340, Abnova™
Shop All Abnova Corporation ProductsDescription
Specifications
Specifications
| Antigen | HIST1H3A |
| Applications | ChIP Assay, Multiplex |
| Classification | Monoclonal |
| Clone | RM340 |
| Conjugate | Unconjugated |
| Description | histone cluster 1, H3a |
| Gene | HIST1H3A |
| Gene Alias | H3/A, H3FA |
| Host Species | Rabbit |
| Immunogen | A trimethyl-peptide corresponding to HIST1H3A (trimethyl K4) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.