Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIST1H4A Rabbit anti-Human, Mouse, Unconjugated, Clone: DEF-8, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20163
Description
HIST1H4A Monoclonal antibody specifically detects HIST1H4A in Human, Mouse samples. It is validated for WB, ICC, IF.Specifications
| HIST1H4A | |
| Monoclonal | |
| Unconjugated | |
| HIST1H4A | |
| Rabbit | |
| 100 μg | |
| 8359 | |
| Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| DEF-8 | |
| histone cluster 1, H4a | |
| H4/a, H4FA | |
| A synthetic peptide corresponding to human HIST1H4A (mono methyl K16) | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction