Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST1H4A, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89998382 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-382 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-382 Supplier Abnova Corporation Supplier No. PAB31333
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen HIST1H4A
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA (1:100-500) Western Blot (1:500) ChIP (1-2 ug/IP) Dot Blot (1:5000) Immunofluorescence (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide, 0.05% proclin 300).
Gene histone cluster 1, H4a
Gene Alias H4/a/H4FA
Gene Symbols HIST1H4A
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to Histone H4, acetylated at lysine 5.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8359
Target Species Human, Mouse
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.