Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIST2H2AA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543692
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543692 Supplier Invitrogen™ Supplier No. PA543692
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PRHLQLAIRN DEELNKLLGK VTIAQGGVLP NIQAVLLPKK TESHHKAKGK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a replication-dependent histone that is a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIST2H2AA3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene H2AC18
Gene Accession No. Q6FI13
Gene Alias H2A; H2A histone family, member O; H2A.2; H2A/O; H2A/q; H2a-615; H2AFO; HIST2H2AA; Hist2h2aa1; HIST2H2AA3; HIST2H2AA4; histone 2, H2aa; histone 2, H2aa3; histone cluster 2, H2aa; histone cluster 2, H2aa1; histone cluster 2, H2aa3; histone H2A type 2-A; Histone H2A.2; Histone H2A/o
Gene Symbols H2AC18
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human HIST2H2AA3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8337
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.