Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIST2H2AC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543691
Description
Peptide sequence: IPRHLQLAIR NDEELNKLLG KVTIAQGGVL PNIQAVLLPK KTESHKAKSK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 92%; Rat: 100%.
Histones are relatively small proteins that contain a high proportion of positively charged amino acids, e.g., arginine and lysine. This positive charge allows histones to bind tightly to the negatively charged DNA. There are five basic types of histone molecules called H1, H2A, H2B, H3, and H4. Histones are abundantly present in virtually all eukaryotic cells.
Specifications
HIST2H2AC | |
Polyclonal | |
Unconjugated | |
H2AC20 | |
H2A; H2A clustered histone 20; H2A histone family, member Q; H2A/q; H2a-613B; H2AC20; H2AFQ; H2A-GL101; Hist2h2ab; HIST2H2AC; histone 2, H2ac; histone cluster 2, H2ac; histone H2A type 2-C; histone H2A/q; Histone H2A-GL101; histone IIa; MGC74460; RGD1560965 | |
Rabbit | |
Protein A | |
RUO | |
8338 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q16777 | |
H2AC20 | |
Synthetic peptide directed towards the middle region of human HIST2H2AC. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction