Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIST2H2BE Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560199
Description
Immunogen sequence: KESYSIYVYK VLKQVHPDTG ISSKAM Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Specifications
HIST2H2BE | |
Polyclonal | |
Unconjugated | |
H2BC21 | |
AV127319; GL105; H2B; H2b 613; H2B histone family, member Q; H2B.1; H2B/q; H2b-613; H2BFQ; H2BGL105; H2BQ; Hist2h2be; HIST2H2BF; histone 2, H2be; histone cluster 2, H2be; histone cluster 2, H2bf; histone H2B type 2-E; Histone H2B type 2-F; Histone H2B.q; histone H2B-GL105; Histone H2Bq; RGD1562346; T25626 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
319190, 8349 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q16778, Q64524 | |
H2BC21 | |
Recombinant protein corresponding to Human HIST2H2BE. Recombinant protein control fragment (Product #RP-104350). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction