Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIST2H2BE Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549266
Description
Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Specifications
HIST2H2BE | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q16778 | |
HIST2H2BE | |
KLH conjugated synthetic peptide between 12-40 amino acids from the N-terminal region of human HIST2H2BE. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
HIST2H2BE | |
Histone H2B type 2-E, Histone H2B-GL105, Histone H2Bq, H2B/q, HIST2H2BE, H2BFQ | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8349 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction