Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST2H3C(27Ac) Mouse anti-Human, Clone:6E7A9, Abnova™

Catalog No. 89333472 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-472 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-472 Supplier Abnova Corporation Supplier No. MAB17151
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of human HIST2H3C(27Ac).

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. This structure consists of approximately 146 bp of DNA wrapped around a nucleosome, an octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is found in a histone cluster on chromosome 1. This gene is one of four histone genes in the cluster that are duplicated; this record represents the telomeric copy. [provided by RefSeq]

Specifications

Antigen HIST2H3C(27Ac)
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 6E7A9
Conjugate Unconjugated
Description histone cluster 2, H3c
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry (1:200-1:1000)
Immunocytochemistry
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene HIST2H3C
Gene Alias H3, H3.2, H3/M, H3F2, H3FM, MGC9629
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acid of human HIST2H3C(27Ac).
Quantity 100 μg
Gene ID (Entrez) 126961
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.