Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histamine dihydrochloride, 99%

Catalog No. AC150625000
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Catalog No. AC150625000

Supplier: Thermo Scientific Chemicals 150625000

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 56-92-8 | C5H11Cl2N3 | 184.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-92-8
Molecular Formula C5H11Cl2N3
Molecular Weight (g/mol) 184.06
MDL Number MFCD00012703
InChI Key PPZMYIBUHIPZOS-UHFFFAOYSA-N
Synonym histamine dihydrochloride, histamine 2hcl, 2-1h-imidazol-4-yl ethanamine dihydrochloride, 1h-imidazole-4-ethanamine dihydrochloride, 1h-imidazole-4-ethanamine, dihydrochloride, peremin, ceplene, 2-1h-imidazol-4-yl ethylamine dihydrochloride, 2-4-imidazolyl ethylamine dihydrochloride, histaminedium dichloride
PubChem CID 5818
IUPAC Name 2-(1H-imidazol-5-yl)ethanamine;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1

Specifications

Melting Point 249.0°C to 252.0°C
Color White
Loss on Drying 0.5% (105°C) max.
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Glass bottle
Quantity 500 g
Beilstein 25315
Merck Index 154756
Solubility Information Solubility in water: freely soluble in water. Other solubilities: soluble in ethanol,freely soluble in methanol
Formula Weight 184.07
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Histamine dihydrochloride
Show More Show Less
Hazard Category Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Respiratory sensitisation (category 1), Skin sensitisation (category 1), Skin corrosion/irritation (category 2)
Hazard Statement Causes serious eye irritation, May cause respiratory irritation, May cause allergy or asthma symptoms or breathing difficulties if inhaled, May cause an allergic skin reaction, Causes skin irritation
EINECSNumber 200-298-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.