Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Histamine, Free Base, ≥97%, Calbiochem™,

Catalog No. 37795GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
37-795-GM 5 g Glass Bottle
37-791-GM 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-795-GM Supplier MilliporeSigma™ Supplier No. 37795GM

Chemical Identifiers

CAS 51-45-6
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.15
MDL Number MFCD00005210,MFCD00128939
InChI Key NTYJJOPFIAHURM-UHFFFAOYSA-N
Synonym histamine, 1h-imidazole-4-ethanamine, 2-4-imidazolyl ethylamine, ergotidine, 2-1h-imidazol-5-yl ethanamine, ergamine, eramin, 5-imidazoleethylamine, theramine, free histamine
PubChem CID 774
ChEBI CHEBI:18295
IUPAC Name 2-(1H-imidazol-5-yl)ethan-1-amine
SMILES NCCC1=CN=CN1

Specifications

Color White
Packaging Glass Bottle
Quantity 5 g
Percent Purity ≥97% by TLC
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.