Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histamine H1R Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP314373
This item is not returnable.
View return policy
Description
Histamine H1R Polyclonal antibody specifically detects Histamine H1R in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Histamine H1R | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| H1R, H1-R, hisH1, histamine receptor H1, subclass H1 | |
| Synthetic 19 amino acid peptide from 3rd cytoplasmic domain of human Histamine H1R. | |
| 50 μg | |
| GPCR, Immune System Diseases, Immunology, Innate Immunity | |
| 3269 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction