Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histamine H1R Antibody (480054), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4726I
Description
Histamine H1R Monoclonal antibody specifically detects Histamine H1R in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Histamine H1R | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
480054 | |
Flow Cytometry, CyTOF-ready | |
H1R, H1-R, hisH1, histamine receptor H1, subclass H1 | |
NS0 mouse myeloma cell line transfected with human Histamine H1 R, Met1-Ser487, Accession # AAH60802 | |
0.1 mL | |
GPCR, Immune System Diseases, Immunology, Innate Immunity | |
3269 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction