Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histamine H1R Antibody (480054), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4726MFV500
Description
Histamine H1R Monoclonal antibody specifically detects Histamine H1R in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| Histamine H1R | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 480054 | |
| Flow Cytometry, CyTOF-ready | |
| H1R, H1-R, hisH1, histamine receptor H1, subclass H1 | |
| NS0 mouse myeloma cell line transfected with human Histamine H1 R, Met1-Ser487, Accession # AAH60802 | |
| 0.1 mL | |
| GPCR, Immune System Diseases, Immunology, Innate Immunity | |
| 3269 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction