Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histamine H3R Antibody (1008944) [Janelia Fluor™ 646], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10200J
Description
Histamine H3R Monoclonal antibody specifically detects Histamine H3R in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Histamine H3R | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
1008944 | |
Flow Cytometry, CyTOF-ready | |
GPCR97G protein-coupled receptor 97, G-protein coupled receptor 97, H3R, HH3R, histamine H3 receptor, histamine receptor H3 | |
Synthetic peptide containing human Histamine H3 R, Accession # Q9Y5N1 | |
0.1 mL | |
GPCR, Immunology, Innate Immunity | |
11255 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction