Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histamine H4R Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Histamine H4R |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Histamine H4R Polyclonal specifically detects Histamine H4R in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Histamine H4R | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
AXOR35H4, GPCR105MGC133027, G-protein coupled receptor 105, GPRv53histamine H4 receptor, H4Rpfi-013, HH4RBG26, histamine receptor H4, Pfi-013, SP9144 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 194-304 of human Histamine H4R (NP_067637.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
GPCR, Immunology, Innate Immunity | |
PBS with 50% glycerol, pH7.3. | |
59340 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title