Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone acetyltransferase KAT2A, Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABE141825UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE141825UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE141825UG Supplier MilliporeSigma Supplier No. ABE141825UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Histone acetyltransferase KAT2A, Cat. No. ABE1418, is a highly specific rabbit polyclonal antibody that detects KAT2A (GCN5) and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Histone acetyltransferase KAT2A (UniProt: Q92830; also known as EC 2.3.1.48, General control of amino acid synthesis protein 5-like 2, Histone acetyltransferase GCN5, HsGCN5, Lysine acetyltransferase 2A, STAF97) is encoded by the KAT2A (also known as GCN5, GCN5L2, HGCN5) gene (Gene ID: 2648) in human. Histone acetyltransferases (HATs) are transcription co-activators recruited by transcriptional activators to target gene promoters, where they enhance the transcription activity by acetylating lysine residues within the N-terminal regions of histones. KAT2A has significant histone acetyltransferase activity with core histones, but not with nucleosome core particles. It is also shown to acetylate non-histone proteins, such as CEBPB. It is also responsible for the acetylation of histone H3 at the gamma-H2A.X harboring nucleosomes at the site of DNA damage, thus establishing a cross-talk between the two histone modifications. It is expressed in all tissues and high expression is observed in ovary. Two isoforms of KAT2A have been described that are produced by alternative splicing. It has three acetyl CoA-binding sites (aa 579-581; 586-592, and 617-620) and an N-acetyltransferase domain (aa 503-656) and a bromo domain (aa 745-815). Histone acetyltransferase KAT2A plays important roles in multiple stages of T cell functions, including development, clonal expansion, and differentiation. Loss of its activity is shown to impair T cell proliferation, IL-2 production, and Th1/Th17, but not Th2 and regulatory T cell differentiation. In case of HIV-1 infection, it is recruited by the viral protein Tat and regulates Tat's transactivating activity and may help inducing chromatin remodeling of proviral genes. (Ref.: (Ref.: Gao, B et al. (2017). J. Immunol. 198(10); 3927-3938).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone acetyltransferase KAT2A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Histone acetyltransferase KAT2A in human pancreas, cerebral cortex, and tonsil tissue sections.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias General control of amino acid synthesis protein 5-like 2;Histone acetyltransferase GCN5;HsGCN5;Lysine acetyltransferase 2A;STAF97
Gene Symbols KAT2A;GCN5;GCN5L2;HGCN5
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal half of human Histone acetyltransferase KAT2A.
Molecular Weight of Antigen ∽94 kDa observed; 93.93 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Histone acetyltransferase KAT2A in human cells. It targets an epitope within 15 amino acids from the C-terminal half.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.