Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone Deacetylase 2/HDAC2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241285
Description
Histone Deacetylase 2/HDAC2 Polyclonal specifically detects Histone Deacetylase 2/HDAC2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
Histone Deacetylase 2/HDAC2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
HDAC2 | |
Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human HDAC2. The immunogen is located within the last 50 amino acids of HDAC2. Amino Acid Squence: KKARIEEDKKETEDKKTD | |
Affinity Purified | |
RUO | |
Primary | |
At least two isoforms of HDAC2 are known to exist; this antibody will detect both isoforms. HDAC2 antibody is predicted to not cross-react with other members of the HDAC family. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 0.5-1 ug/ml, ELISA, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence 20 ug/ml | |
EC 3.5.1.98, HD2, histone deacetylase 2, RPD3, transcriptional regulator homolog RPD3, YAF1, YY1-associated factor 1 | |
Rabbit | |
52 kDa | |
0.1 mg | |
Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases, Stem Cell Markers | |
3066 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction