Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone Deacetylase 2/HDAC2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP144095 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144095 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144095 Supplier Novus Biologicals Supplier No. NBP144095
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone Deacetylase 2/HDAC2 Polyclonal specifically detects Histone Deacetylase 2/HDAC2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Histone Deacetylase 2/HDAC2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA 1:20000-1:60000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 30% Glycerol with No Preservative
Gene Accession No. NP_001518
Gene Alias EC 3.5.1.98, HD2, histone deacetylase 2, RPD3, transcriptional regulator homolog RPD3, YAF1, YY1-associated factor 1
Gene Symbols HDAC2
Host Species Rabbit
Immunogen Histone Deacetylase 2/Histone Deacetylase 2/HDAC2 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the near C-terminal region of human Histone Deacetylase 2/HDAC2. (Uniprot: Q92769)
Molecular Weight of Antigen 55.36 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3066
Test Specificity This antibody is specific to HDAC2
Reconstitution Reconstitute with 185 ml of water to desired concentration and for immediate usage.
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.