Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone Deacetylase 2/HDAC2 Antibody (OTI7E10), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245949
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
Histone Deacetylase 2/HDAC2 Monoclonal specifically detects Histone Deacetylase 2/HDAC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Histone Deacetylase 2/HDAC2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
EC 3.5.1.98, HD2, histone deacetylase 2, RPD3, transcriptional regulator homolog RPD3, YAF1, YY1-associated factor 1 | |
Mouse | |
55.8 kDa | |
0.1 mL | |
Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases, Stem Cell Markers | |
3066 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI7E10 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
HDAC2 | |
Human recombinant protein fragment corresponding to amino acids 415-488 of human HDAC2(NP_001518) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction