Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone Deacetylase 2/HDAC2 Rabbit anti-Human, Mouse, Rat, Clone: S06-4E4, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | Histone Deacetylase 2/HDAC2 |
---|---|
Clone | S06-4E4 |
Dilution | Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence 20 ug/ml |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Monoclonal |
Description
Histone Deacetylase 2/HDAC2 Monoclonal antibody specifically detects Histone Deacetylase 2/HDAC2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ImmunofluorescenceSpecifications
Histone Deacetylase 2/HDAC2 | |
Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence 20 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 3.5.1.98, HD2, histone deacetylase 2, RPD3, transcriptional regulator homolog RPD3, YAF1, YY1-associated factor 1 | |
Recombinant protein of human Histone Deacetylase 2/HDAC2 (Uniprot#: Q92769) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S06-4E4 | |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Chromatin Research, Epigenetics, Stem Cell Markers | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
3066 | |
IgG | |
Affinity purified | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title