Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone Deacetylase 4/HDAC4 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222151G
Description
Histone Deacetylase 4/HDAC4 Polyclonal specifically detects Histone Deacetylase 4/HDAC4 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Histone Deacetylase 4/HDAC4 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
HA6116, HD4EC 3.5.1.98, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HDAC4 | |
A synthetic peptide made to a C-terminal portion of the human HDAC4 protein (between residues 1000-1084). [UniProt P56524] | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Phospho Specific | |
Human, Mouse, Rat, Primate | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction