Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone Deacetylase 8/HDAC8 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298969MFV450
Description
Histone Deacetylase 8/HDAC8 Polyclonal antibody specifically detects Histone Deacetylase 8/HDAC8 in Human samples. It is validated for Western BlotSpecifications
Histone Deacetylase 8/HDAC8 | |
Polyclonal | |
Western Blot | |
EC 3.5.1.98, HD8, HDACL1, histone deacetylase 8, histone deacetylase-like 1, RPD3 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Histone Deacetylase 8/HDAC8. | |
0.1 mL | |
Breast Cancer, Cancer, Chromatin Research, Epigenetics, Phospho Specific | |
55869 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction