Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone Deacetylase 8/HDAC8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$244.50 - $536.00
Specifications
Antigen | Histone Deacetylase 8/HDAC8 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Histone Deacetylase 8/HDAC8 Polyclonal specifically detects Histone Deacetylase 8/HDAC8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
Histone Deacetylase 8/HDAC8 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 3.5.1.98, HD8, HDACL1, histone deacetylase 8, histone deacetylase-like 1, RPD3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-377 of human Histone Deacetylase 8/HDAC8 (NP_060956.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Chromatin Research, Epigenetics, Phospho Specific | |
PBS with 50% glycerol, pH7.3. | |
55869 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title