Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Histone H1.0 Recombinant Rabbit Monoclonal Antibody (5E3Q8)
GREENER_CHOICE

Catalog No. PIMA535484
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535484 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535484 Supplier Invitrogen™ Supplier No. MA535484
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MTENSTSAPA AKPKRAKASK KSTDHPKYSD MIVAAIQAEK NRAGSSRQSI QKYIKSHYKV GENADSQIKL SIKRLVTTGV LKQTKGVGAS GSFRLAKSDE.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a replication-independent histone that is a member of the histone H1 family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H1.0
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 5E3Q8
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene H1-0
Gene Accession No. P07305, P10922
Gene Alias D130017D06Rik; H1 histone family member 0; H1 histone family, member 0; H1(0); H1.0, H1(0), H1-0; H10; H1-0 histone; H1f0; H1fv; histone H1'; Histone H1(0); histone H1.0; Histone H1.0, N-terminally processed; Histone H1-0; MyD196
Gene Symbols H1-0, H1F0
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Histone H10 (P07305).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14958, 3005
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.