Learn More
Invitrogen™ Histone H1.2 Polyclonal Antibody

Description
Immunogen sequence: KKAGATAGKR KASGPPVSEL ITKAVAA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 85%.
Specifications
Specifications
| Antigen | Histone H1.2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | H1-2 |
| Gene Accession No. | P16403 |
| Gene Alias | 0610008C09Rik; CTL-1; H1 histone family, member 2; H1 VAR.1; H1.2; H1.2 linker histone, cluster member; H1-2; H1C; H1f2; H1s-1; H1var1; His1a; Hist1h1c; histone 1, H1c; histone cluster 1, H1c; histone H1; histone H1.1; histone H1.2; Histone H1c; histone H1d; Histone H1s-1; LOC684681; MGC3992; similar to Histone H1.2 (H1 VAR.1) (H1c) |
| Gene Symbols | H1-2, HIST1H1C |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.